Learn More
Abnova™ STK33 Recombinant Protein
Recombinant protein for STK33 (human) gene
Brand: Abnova™ H00065975-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Human STK33 full-length ORF ( AAH31231, 1 a.a. - 514 a.a.) recombinant protein with GST-tag at N-terminal
- serine/threonine kinase 33
- Theoretical molecular weight: 82.28kDa
- Preparation: in vitro wheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer
Sequence: MADSGLDKKSTKCPDCSSASQKDVLCVCSSKTRVPPVLVVEMSQTSSIGSAESLISLERKKEKNINRDITSRKDLPSRTSNVERKASQQQWGRGNFTEGKVPHIRIENGAAIEEIYTFGRILGKGSFGIVIEATDKETETKWAIKKVNKEKAGSSAVKLLEREVNILKSVKHEHIIHLEQVFETPKKMYLVM ELCEDGELKEILDRKGHFSENETRWIIQSLASAIAYLHNNDIVHRDLKLENIMVKSSLIDDNNEINLNIKVTDFGLAVKKQSRSEAMLQATCGTPIYMAPEVISAHDYSQQCDIWSIGVVMYMLLRGEPPFLASSEGKLFELIRKGELHFENAVWNSISDCAKSVLKQLMKVDPAHRITAKELLDNQWLTGNKLSSVRPTNVLEMMKEWKNNPESVEENTTEEKNKPSTEEKLKSYQPWGNVPETNYTSDEEEEKQSTTYEKQFPATSKDNFDMCSSSFTSSKLLPAEIKGEMEKTPVTPSQGTATKYPAKSGALSRTKKKL
Best use within three months from the date of receipt.
ELISA, western blotting (recombinant protein), antibody production, protein array
Specifications
AAH31231 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
82.28 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MADSGLDKKSTKCPDCSSASQKDVLCVCSSKTRVPPVLVVEMSQTSSIGSAESLISLERKKEKNINRDITSRKDLPSRTSNVERKASQQQWGRGNFTEGKVPHIRIENGAAIEEIYTFGRILGKGSFGIVIEATDKETETKWAIKKVNKEKAGSSAVKLLEREVNILKSVKHEHIIHLEQVFETPKKMYLVMELCEDGELKEILDRKGHFSENETRWIIQSLASAIAYLHNNDIVHRDLKLENIMVKSSLIDDNNEINLNIKVTDFGLAVKKQSRSEAMLQATCGTPIYMAPEVISAHDYSQQCDIWSIGVVMYMLLRGEPPFLASSEGKLFELIRKGELHFENAVWNSISDCAKSVLKQLMKVDPAHRITAKELLDNQWLTGNKLSSVRPTNVLEMMKEWKNNPESVEENTTEEKNKPSTEEKLKSYQPWGNVPETNYTSDEEEEKQSTTYEKQFPATSKDNFDMCSSSFTSSKLLPAEIKGEMEKTPVTPSQGTATKYPAKSGALSRTKKKL | |
STK33 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Kinase Assay, Western Blot | |
65975 | |
STK33 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
STK33 | |
Wheat Germ (in vitro) | |
GST |