Learn More
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers
Product Code. 15133007 Shop All Fisherbrand Products
Click to view available options
Size:
305mm
457mm
610mm
762mm
Includes:
Propellor, 108mm
Propellor, 63.5mm
Unit Size:
Each
Product Code. 15133007
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Excellent at producing axial flows in vessels from top to bottom. Fisherbrand™ Stainless Steel Stirring Shafts with Propellers are for use with overhead stirrer units.
  • Available in a range of sizes
  • Resistant to corrosion
Specifications
Includes Propellor, 63.5mm
Quantity 1 Each
Size 762mm
Type Stirring Shaft