Learn More
Bohlender Stirrer Bearings NS 19/26 6mm GL 18
Brand:  Bohlender C424-04
Stirrer Bearings NS 19/26 6mm GL 18
Product Code. 10023321
  • ONLINE EXCLUSIVE PRICE
    49.35€
    41.68€ / Pack of 1
    Save 7.67€
    15% Off
Estimated Shipment: 22-10-2026