Learn More
Cole-Parmer™ Stir Bars
Product Code. 12789038
Click to view available options
Length (Metric):
10 mm
12 mm
Diameter (Metric):
4.5 mm
6 mm
Shape:
Cylinder
Oval
Unit Size:
Pack of 10
Product Code. 12789038
Brand: Cole-Parmer™ 3663098
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Stir Bars suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Specifications
Diameter (English) 0.24 in.
For Use With (Application) For Stem Integrity
For Use With (Equipment) Integrity 10, RS-200, and RS-250 Reaction Stations
Length (English) 0.4 in.
Diameter (Metric) 6 mm
Length (Metric) 10 mm
Shape Oval
Type Magnetic Stir Bar