Learn More
STF-118804, TRC
Specifications
Specifications
| Chemical Name or Material | 4-[5-Methyl-4-[((4-methylphenyl)sulfonyl)methyl]-2-oxazolyl]-N-(3-pyridinylmethyl)benzamide |
| CAS | 894187-61-2 |
| Molecular Formula | C25H23N3O4S |
| Quantity | 5 mg |
| Synonym | STF-118804, 4-[5-Methyl-4-[[(4-methylphenyl)sulfonyl]methyl]-2-oxazolyl]-N-(3-pyridinylmethyl)benzamide |
| InChI Formula | InChI=1S/C25H23N3O4S/c1-17-5-11-22(12-6-17)33(30,31)16-23-18(2)32-25(28-23)21-9-7-20(8-10-21)24(29)27-15-19-4-3-13-26-14-19/h3-14H,15-16H2,1-2H3,(H,27,29) |
| SMILES | Cc1ccc(cc1)S(=O)(=O)Cc2nc(oc2C)c3ccc(cc3)C(=O)NCc4cccnc4 |
| IUPAC Name | 4-[5-methyl-4-[(4-methylphenyl)sulfonylmethyl]-1,3-oxazol-2-yl]-N-(pyridin-3-ylmethyl)benzamide |
| Molecular Weight (g/mol) | 461.53 |
| Formula Weight | 461.1409 |