Learn More
Sampling Systems™ SteriWare™ Disposable Volumetric Scoop
BIOPHARMA
Product Code. p-7138880 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
1 mL
1.23 mL
2.46 mL
5 mL
10 mL
20 mL
Length (Metric):
153 mm
155 mm
160 mm
165 mm
171 mm
180 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 12734948
Brand: Sampling Systems™ 8640H01S
Capable of being used straight from the bag with its individually bagged design. Sampling Systems™ SteriWare™ Disposable Volumetric Scoop allows small volumes to be accurately measured.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Available in a variety of sizes
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contaminaton
  • No cleaning required
Specifications
Sterility Sterile
Capacity (Metric) 1 mL
Disposable Single-use
Product Type Volumetric Scoop
Diameter (Metric) Outer 18 mm
Material HDPE
Length (Metric) 153 mm
These Volumetric Spoons are not certified measuring devices. User verification required.