Learn More
Sampling Systems™ SteriWare™ Disposable Slot Sampler
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
100 mL
170 mL
500 mL
Length (Metric):
620 mm
970 mm
1000 mm
Diameter (Metric):
25 mm
40 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
Beschreibung
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Suitable for use in ATEX areas
- Push the sampler into the product, ensuring the sampler is in the closed position
- After leaving the sampler for several seconds to allow powder to enter, rotate handle to close the sampler
- Deposit the sample into a suitable container
- No cross contaminatin
- No cleaning required
Spezifikation
Spezifikation
| Product Type | Disposable Slot Sampler |
| Material | HDPE |
| Includes | 20 per pack |
| Capacity (Metric) | 170 mL |
| Sterility | Non-sterile |
| Diameter (Metric) | 25 mm |
| Length (Metric) | 970 mm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur