Learn More
Sampling Systems™ SteriWare™ Disposable Powder Bottle
Shop All Sampling Systems Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
250 mL
500 mL
Sterility:
Non-sterile
Sterile
missing translation for 'unitSize'
Pack of 37
Pack of 50
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Available in 2 sizes
- Powder tight seal
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contaminaton
- No cleaning required
Specifications
Specifications
| Product Type | Bottle |
| Material | Polypropylene |
| Includes | Individually Bagged |
| Capacity (Metric) | 500 mL |
| Sterility | Non-sterile |
| Diameter (Metric) | 106 mm |
| Length (Metric) | 108 mm |