Learn More
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette
BIOPHARMA
Product Code. 16592170 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
225 mL
Unit Size:
Pack of 37
Product Code. 16592170
Brand: Sampling Systems™ 8051H1100
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette is both quick and easy to use and is ideal for taking samples from low viscosity liquids.
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Highly visible color coded handle
  • Press thumb onto blue end of the tube until lowered to the correct sampling depth
  • At the required depth raise thumb until levels equalize
  • Press the thumb onto tube to seal in the sample
  • No cross contamination
  • No cleaning required
Specifications
Material HDPE
Includes Individually Bagged
Capacity (Metric) 225 mL
Sterility Non-sterile
Diameter (Metric) Outer 21 mm
Length (Metric) 1100 mm