Learn More
Sampling Systems™ SteriWare™ Capsule Counting Trays
Shop All Sampling Systems Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
Description
- Our capsule counting trays makes accurate sorting and counting easy.
- They are made from robust, shatterproof PS.
- They are suitable for use with all capsules from Size 2 to Size 00.
- Use straight from the heat sealed PE bag.
- They are manufactured and packed under cleanroom conditions.
- Made from white food grade PS.
- Conforms to the latest FDA and EU requirements.
- BSE/TSE free.
- Either sterile or non sterile.
Specifications
Specifications
| Height (Metric) | 12 mm |
| Length (Metric) | 260 mm |
| Material | Polypropylene |
| Width (Metric) | 130 mm |
| Sterility | Sterile |
| Packaging Type | Bag |