Learn More
Sartorius Sterisart™ Easy Configuration Software
Product Code. 17288720 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Sterisart Universal Pump
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17288720 Sterisart Universal Pump Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17288720 Supplier Sartorius Supplier No. 1Z0004
  • ONLINE EXCLUSIVE PRICE
    5890.00€
    5224.49€ / Pack of 1
    Save 665.51€
    11% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Sterisart™ Easy Configuration Software is drag and drop software designed to increase process reliability for the sterility testing of sterile pharmaceuticals.
  • Step-by-step guidance through each sterility test programmed, such as for USP 71 or Eu. Ph. 2.6.1
  • FDA 21 CFR Part 11 compliant electronic records
  • User account management, audit trail and password protection
  • Scanning, saving and printing of process-relevant data and records on materials
TRUSTED_SUSTAINABILITY
Specifications
Type Automate Online Configuration Tool
For Use With (Equipment) Sterisart Universal Pump