missing translation for 'onlineSavingsMsg'
Learn More

BD Sterility Bottles

Product Code. 11713593
Click to view available options
Quantity:
100 mL
300 mL
Color:
Beige
Clear
Light Yellow
Media Type:
Fluid A
Fluid D
Trypticase Soy Broth
Unit Size:
1 L
3 L
Pack of 10
This item is not returnable. View return policy

Product Code. 11713593

Brand: BD 299107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Certificate of Analysis

Glass sterility bottles available with a variety of media

  • Transparent labels for easy viewing and identification of medium
  • Labels are laminated acetate to resist alcohol or chlorine treatment for cleanroom use
  • Prepared bottles for sterility testing and microbial limits

Wide-Mouth Bottles

  • Openings are 48mm in diameter to accommodate bulky samples

Order Info

Supplied in packs of 10 bottles

Compliance

Conform to harmonized USP requirements.

TRUSTED_SUSTAINABILITY

Specifications

Cap Type Flip-top
Certifications/Compliance USP
Color Beige
Form Liquid
Manufacturing Origin United States
Media Type Trypticase Soy Broth
Fill Volume 100 mL
For Use With (Application) Sterility testing and microbial limits
Includes Certificate of Analysis
Packaging Type Bottle
Quantity 100 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.