Learn More
Thermo Scientific™ Sterilin™ Cuvets
Product Code. 12751548
Click to view available options
Capacity (Metric):
1.5 to 3.0 mL
2.5 to 4.5 mL
Material:
PMMA
Polystyrene
Description:
Macro cuvette
Semi-micro cuvette
Unit Size:
Pack of 500
Product Code. 12751548
Brand: Thermo Scientific™ 221S
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Experience top optical quality in spectrophotometry with easy-to-use, polystyrene or acrylic Thermo Scientific™ Sterilin™ Cuvets.
Experience top optical quality in spectrophotometry with easy-to-use, polystyrene or acrylic Thermo Scientific™ Sterilin™ Cuvets.
  • Choice of material: optical quality polystyrene for use at visible wavelengths (340 to 900nm) or optical quality acrylic (PMMA) for use at ultraviolet wavelengths (300 to 900nm)
  • Each case contains cuvettes from the same mold cavity to ensure minimal dimensional variation
  • Standard absorption variation between cuvettes is better than ±0.005 absorbency units
  • Two sizes available—semi-micro cuvette and macro cuvette
  • Standard pathlength of 10mm
  • Frosted sides on the cuvette provide an ideal labeling and handling area
  • Recessed windows reduce the risk of scratching during use
  • For ease of use an arrow feature indicates the direction of light transmission
  • Packed in expanded polystyrene (EPS) foam trays for optimum protection ensuring scratchfree product at the time of use
Ordering Information
  • Not available in all geographic regions
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
Type Semi-micro Cuvette
Description Semi-micro cuvette
Capacity (Metric) 1.5 to 3.0 mL
Material Polystyrene
Quantity 100 pk/ 500 cs
Path Length 10 mm
Wavelength Range 340 to 900 nm
Height (Metric) 22 mm
For Use With (Equipment) Spectrophotometers, Photometers
Width (Metric) 4.5 mm
Show More Show Less
Note: cuvettes should not be used for long-term storage of samples. When using Hydrochloric acid, as the instrument can come under attack from the acid fumes, it is recommended that you use sealing film on the cuvette.