Learn More
Thermo Scientific™ Sterilin™ BenchGuard
GREENER_CHOICE
Product Code. 11952568
Click to view available options
Description:
BenchGuard
BenchGuard Extra
Absorbancy:
400 mL/m2
800 mL/m2
Quantity:
1 x 50m Roll
100 x Sheets
50 x Sheets
Unit Size:
50 sheets
Each
Pack of 100
Product Code. 11952568
Brand: Thermo Scientific™ BG60/100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Protect benches and other surfaces against liquid spills with the highly absorbent Thermo Scientific™ Sterilin™ BenchGuard.
Protect benches and other surfaces against liquid spills with the highly absorbent Thermo Scientific™ Sterilin™ BenchGuard. Absorbent liner also provides a cushion for glassware when accidentally dropped on benchtop.
  • Available in both 400mL/m2 and 800mL/m2absorption capacities
  • Plastic coated on one side to prevent soak through
  • Available in roll or sheet form
  • Rolls supplied in easy to use dispenser packs
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bench Guard
Absorbancy 400 mL/m2
Length (English) 1.607612 ft.
Length (Metric) 0.6 m
Width (English) 1.607612 ft.
Width (Metric) 0.49 m
Description BenchGuard
Quantity 100 x Sheets