Learn More
Bel-Art™ Sterileware™ Cross-Sectional Liquid Samplers
Product Code. p-7214229 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
80 mL
225 mL
Unit Size:
Pack of 20
Product Code. 16806926
Brand: Bel-Art™ F378740450
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The integrated valve of this sterile tool closes as it is pushed against a container bottom securing the sample for withdrawal. Sample can then be poured from the top of the tool into your sample container.
  • Provides effective cross-sectional sampling of liquids and light creams
  • Reaches bottom of container while keeping hand clear of container top
  • Diameter allows for use through drum bunghole or other small openings
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) samplers heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Product Type Liquid Sampler
Material HDPE
Autoclavable Not Autoclavable
Capacity (English) 2.75 oz.
Capacity (Metric) 80 mL
Quantity 20/Pk.
Diameter (Metric) Outer 2.1 cm
Length (Metric) 45 cm