Learn More
Keystone Cleanroom Products Sterile Zip Lock Bag with Slide Closure
Shop All Keystone Cleanroom Products Products
Click to view available options
Length (English):
12 in.
15 in.
No. per Pack:
2 Pk.
20 Pk.
Unit Size:
Pack of 250
Pack of 400
Pack of 500
Description
- Pre-sterilized and packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitisation agents
- The bags are used for the removal and transport of media plates from the aseptic area to QC laboratories
- Manufactured and packaged in an ISO certified cleanroom
- Tear and puncture resistant 2.7 mill low density polyethylene
- Each bag has an integral zip seal with the slide lock closure system
- Two zip-lock bags are double packaged in Tyvek/film chevron peel pouches and then inside a third, sealed poly bag layer
- Labelled with lot number, date of manufacture and expiration date
- Validated sterile to a 10-6 SAL
- Certificates of conformance and sterility sent with each shipment
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Closure Type | Press Lock |
| Disposable | Single-use |
| For Use With (Application) | Transport EM Media Plates |
| Material | LDPE |
| Sterility | Sterile |
| Latex Free | Yes |
| No. per Pack | 2 Pk. |
| Sterilization Method | Gamma |
| Thickness (English) | 2.7 mil |
| Show More |