Learn More
Fisherbrand™ Sterile Rodac Bowls
Product Code. 15573790
Click to view available options
Diameter (Metric):
55 mm
Unit Size:
Pack of 500
Product Code. 15573790
Brand: Fisherbrand™ 200218
Prepared, sterile Rodac plates for microbiological testing of all surfaces. Fisherbrand™ Sterile Rodac Bowls provide a simple and quick method to check all kinds of surfaces for microbiological contamination and hygienic status.
  • Rodac: Replicate organism detection and counting.
  • Sterile, prepared plates for surface testing.
Specifications
Sterility Sterile
Type Rodac Plate
Quantity 400 Pack
Diameter (Metric) 55 mm