Learn More
Fisherbrand™ Sterile Polyethylene Sampling Bags
Product Code. 11532783 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
540 mL
720 mL
2250 mL
3500 mL
Includes:
No Writing Area
Writing Area
Unit Size:
Pack of 1000
Product Code. 11532783
Designed with a twirl round metal wire closure system and puncture proof safety tabs. Fisherbrand™ Sterile Polyethylene Sampling Bags are leakproof when sealed.
  • Manufactured with pliant, highly resistant virgin polyethylene 
  • Sterile transparent polyethylene bags without side seals
  • Choose clear or white marking area for written notations
  • Convenient pull tabs to open bag without touching the edges
  • Ideal for use with solid, semi-solid or liquid samples
  • For use in all laboratories where samples need to be collected into a sterile environment.
Specifications
Capacity (Metric) 540 mL
Color Clear
For Use With (Application) Use in all laboratories where samples need to be collected into a sterile environment
Includes Writing Area
Material Polyethylene
Sterility Sterile
Thickness (Metric) 63 μm
Quantity 1000 Pk.
Dimensions (L x W) 229 x 114 mm