Learn More
Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Product Code. 13356048 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
30 μL
Tip Style:
Low Retention
Unit Size:
Pack of 3840
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Tip Style Unit Size
13356048 30 μL Low Retention Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13356048 Supplier Thermo Scientific™ Supplier No. 965261
Estimated Shipment: 28-10-2026
Increase performance through the superior tip straightness and low %CV with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems.
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Filtered
Sterility Sterile
Tip Style Low Retention
For Use With (Equipment) Beckman Coulter Liquid Handling Systems