Learn More
Greiner Bio-One Sterile Inoculation Loop
Product Code. p-8000233
Change view
Click to view available options
Color:
Blue
White
Yellow
Quantity:
2000/Pk.
600/Pk.
Unit Size:
Pack of 2000
Pack of 600
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
10659021 White 2000/Pk. Pack of 2000
10707682 Blue 600/Pk. Pack of 600
10342651 Yellow 600/Pk. Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10659021 Supplier Greiner Bio-One Supplier No. 731101
Only null left
Add to basket
Add to basket
Transfers inoculum for streaking. Greiner Bio-One™ Sterile Inoculation Loops are ideal for retrieving an inoculum from a culture of microorganisms.
  • Flexible loops for easier collection and inoculation
  • Sterilized by irradiation
  • For research only; not for use in diagnostic procedures, unless otherwise indicated
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 1 μL
Length (Metric) 200 mm
Sterility Sterile
Color White
Quantity 2000/Pk.
For Use With (Application) Research use