Learn More
Fisherbrand™ Sterile Graduated Transfer Pipets
Product Code. 13409118 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 mL
3 mL
Length (Metric):
150 mm
155 mm
Volume (Metric) Dispense:
33 μL
40 μL
Packaging Type:
25 inner packs of 20
50 inner packs of 10
Individually Wrapped
Unit Size:
Pack of 500
Product Code. 13409118
Edge
Made up of low density polyethylene.
  • Transparent in color
  • Contains graduations
  • Sterile
Specifications
Material LDPE
Drops/mL 25
Graduated Yes
Graduations 0.5 mL
Quantity 500/Cs.
Volume (Metric) Dispense 40 μL
Packaging Type 25 inner packs of 20
Volume (Metric) 3 mL
Length (Metric) 155 mm
Type Transfer Pipet