Learn More
Texwipe™ Sterile Dry Collection and Transport System
Product Code. 17166199 Shop All Texwipe Products
Click to view available options
Tip Material:
Cotton
Polyester
Length (Metric) Head:
17mm
18.2mm
Width (Metric) Head:
4.6mm
7mm
Unit Size:
Each
Pack of 500
Product Code. 17166199
Brand: Texwipe™ STX764T
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Sterile ready-to-use units for the collection and transport of bacteriological specimens.
Designed to maintain integrity of both sample collection and sample transport to the lab.
  • Constructed from 100% USP-grade spun cotton or 100% USP-grade spun polyester
  • Swabs are bonded securely to a flexible polystyrene shaft by an aqueous-based adhesive
  • 155mm (6.1 in.)long swabs enable access to hard-to-reach sampling sites
  • Medical-grade LDPE molded cap securely holds handle for safety and sterility while sampling
  • Tamper evident seal ensures sterility at point-of-use
  • 1mm thick medical-grade polypropylene tube with seamless molded round-bottom design
  • Cap rings ensure tight seal and protect against contamination during transport
  • Packaging labeled for global use
  • Gamma irradiated to a Sterility Assurance Level of 10-6 in accordance with ANSI/AAMI/ISO 11137
  • Lot code and expiration date clearly marked on each tube
  • Certificates of Irradiation and Compliance attached to each case confirm irradiation dose met specifications
Order Info
1 swab/tube; 100 tubes/box; 5 boxes/case
Specifications
Certifications/Compliance Certificates of Irradiation and Compliance
For Use With (Application) Diagnostic sampling, Environmental monitoring, DNA sampling, Surface sampling
Handle Color White
Product Type Cleanroom Swab
Length (English) Head 0.669 in.
Length (English) Handle 5.276 in.
Length (Metric) Handle 134 mm
Length (Metric) Head 17mm
Width (English) Head 0.181 in.
Width (Metric) Head 4.6mm
Adhesive Type Aqueous-based
Handle Material Polystyrene
Tip Material Polyester