Learn More
Merck Stericup™ Quick Release Bottle Top Receiver Flasks
Product Code. 15987996 Shop All Merck Products
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Product Code. 15987996
Brand: Merck S200B10RE
Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.
Steritop™ bottle top filter units provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33 mm) or wide-mouth (45 mm) bottles.
A small tab inside the funnel secures an optional prefilter in-place during filtration.
For added protection against contamination, the threads for bottle attachment have been slightly recessed in case the funnel accidentally touches a work surface.
TRUSTED_SUSTAINABILITY
Specifications
Outlet Fitting 45 mm
Volume (Metric) 1000 mL