Learn More
Megro™ Steri-Roll
Product Code. 10269073
Change view
Click to view available options
Dimensions (L x W):
200 m x 400 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10269073 200 m x 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10269073 Supplier Megro™ Supplier No. 320400
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Suitable containers for packing different items for steam or gas sterilization. Megro™ Steri-Roll features 3 sterilization indicators for steam, EO gas, and formaldehyde.
  • Suitable for sterilzation of clothes, trays, compresses, and medical instruments.
  • Suitable for sterilization by steam or gas (EO or FO).
  • Indicators for all steam or gas (EO or FO).
  • PET foil and PP laminate construction.
  • Unsuitable for gamma, hot-air, and hydrogen peroxide sterilization.
Specifications
Certifications/Compliance ISO 11607/EN 868
For Use With (Application) Sterilization of clothes, trays, compresses, medical instruments
Material PET, Polypropylene
Quantity 1 Roll
Sterilization Method EO, FO, Gamma irradiation
Dimensions (L x W) 200 m x 400 mm
Length (Metric) 200 m
Width (Metric) 400 mm