Learn More
Zeiss™ SteREO Discovery.V8 Body
Description
SteREO Discovery.V8 microscope bodyApochromatic stereo telescope system 76mm mount diameter 8:1 zoom (1.0x ... 8.0x) Manually operable on both sides Switchable click stops at 1x-1.25x-1.6x-2x-2.5x-3.2x-4x-5x-6.3x-8x
Specifications
Specifications
| Type | Stereo Discovery Microscope Body |
| For Use With (Application) | developmental biology and quality assurance applications |
| For Use With (Equipment) | SteREO Discovery.V8 |
| Magnification Power | 1 to 8X |
| Quantity | 1 Ea. |
| Size | Mount: 76 mm dia. |