Learn More
R&D Systems™ StemXVivo Human Osteogenic Supplement, 12.5 mL
Product Code. 18462050
Change view
Click to view available options
Quantity:
12.5 mL
Unit Size:
12.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18462050 12.5 mL 12.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18462050 Supplier R&D Systems™ Supplier No. CCM008
  • ONLINE EXCLUSIVE PRICE
    247.00€
    233.10€ / 12.5mL
    Save 13.90€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Media supplement to induce the differentiation of human MSCs into osteocytes
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into osteocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Contains high quality differentiation factors to drive reproducible and efficient MSC osteogenesis
  • Is defined to reduce unwanted experimental variability
  • Has been developed and optimized using MSCs
TRUSTED_SUSTAINABILITY
Specifications
Quantity 12.5 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.