Learn More
R&D Systems™ StemXVivo Human/Mouse Chondrogenic Supplement, 0.5 mL
Product Code. 18401570
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18401570 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18401570 Supplier R&D Systems™ Supplier No. CCM006
  • ONLINE EXCLUSIVE PRICE
    247.00€
    233.10€ / 0.5mL
    Save 13.90€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Media supplement to induce the differentiation of MSCs into chondrocytes
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into chondrocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Contains high quality differentiation factors to drive reproducible and efficient MSC chondrogenesis
  • Is defined to reduce unwanted experimental variability.
  • Has been developed and optimized using human and mouse MSCs
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.5 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.