Learn More
R&D Systems™ StemXVivo Hepatocyte Differentiation Kit
Click to view available options
Quantity:
1 Kit
missing translation for 'unitSize'
Each
Beschreibung
- Optimized components ensure highly enriched populations of hepatocyte-like cells
- Reproducible differentiation protocols translate into cost and time savings
- Maximizes workflow efficiency by standardizing hepatocyte differentiation
- Qualified for hepatotoxic drug and small molecule screening
Spezifikation
Spezifikation
| Includes |
|
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| For Use With (Application) | For differentiation of pluripotent stem cell into hepatocytes |
| Product Type | StemXVivo Hepatocyte Differentiation Kit |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur