missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies StemSpan™ SFEM II
Description
- StemSpan™ Serum-Free Expansion Medium II (SFEM II) is a modified version of StemSpan™ SFEM
- It has been developed for the in vitro culture and expansion of human hematopoietic cells
- This medium contains pre-tested bovine serum albumin, insulin, transferrin, and other supplements in Iscove's MDM
- Recombinant hematopoietic growth factors, required for the optimal growth and expansion of hematopoietic cells, have not been added to StemSpan™ SFEM II
- This allows users the flexibility to prepare medium that meets their requirements
- Using appropriate cytokines (e.g. StemSpan™ CC100, StemSpan™ CC110, or StemSpan™ CD34+ Expansion Supplement), StemSpan™ SFEM II can be used for the expansion of total nucleated cells and CD34+ cells from cord blood, bone marrow, or other cell sources
- StemSpan™ SFEM II can also be used to expand and differentiate lineage-committed progenitor cells to generate erythroblasts, granulocytes, monocytes, or megakaryocytes when used with StemSpan™ Erythroid Expansion Supplement (Catalog #02692), StemSpan™ Myeloid Expansion Supplement (Catalog #02693), StemSpan™ Myeloid Expansion Supplement II (Catalog #02694), or StemSpan™ Megakaryocyte Expansion Supplement (Catalog #02696), respectively
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Expansion |
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?