missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies StemSpan™ Lymphoid Progenitor Expansion Supplement (10X)

Product Code. 17149961
Encompass
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 17149961

Brand: STEMCELL Technologies 09915

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Supplement for the expansion and differentiation of lymphoid progenitor cells

  • StemSpan™ NK Cell Generation Kit has been developed to differentiate CD34+ cells isolated from cord blood (CB) and bone marrow (BM) to natural killer (NK) cells, without use of stromal cells
  • This kit typically promotes the expansion of thousands of CD56+ NK cells in cultures initiated with CD34+ human CB cells
  • StemSpan™ Lymphoid Progenitor Expansion Supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion and differentiation of CD34+ cells isolated from human CB and BM samples to lymphoid progenitor cells when used in combination with StemSpan™ SFEM II medium, and on plates coated with StemSpan™ Lymphoid Differentiation Coating Material (100X)
  • Subsequently, StemSpan™ NK Cell Differentiation Supplement (100X) enables differentiation of lymphoid progenitor cells to NK cells

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, NK Cells
For Use With (Application) Cell Culture, Differentiation, Expansion
Quantity 5 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.