Learn More
STEMCELL Technologies StemSpan™ Leukemic Cell Culture Kit
Product Code. 17119961
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17119961 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17119961 Supplier STEMCELL Technologies Supplier No. 09720
 Restricted Item
Estimated Shipment: 25-11-2026
For culture, expansion, and drug screening of chronic and acute myeloid leukemia cells
  • StemSpan™ Leukemic Cell Culture Kit has been developed for the in vitro culture and expansion of malignant cells and has been tested on chronic myeloid leukemia (CML) and acute myeloid leukemia (AML) samples
  • This optimized protocol allows users to expand, culture, and use malignant cells for drug screening
  • StemSpan™ Leukemic Cell Culture Kit includes the serum-free medium SFEM II (Catalog #09605), StemSpan™ CD34+ Expansion Supplement (10X; Catalog #02691), and small molecule UM729 (Catalog #72332)
Specifications
Cell Type Cancer Cells and Cell Lines, Hematopoietic Stem and Progenitor Cells, Leukemia and Lymphoma Cells
For Use With (Application) Cell Culture; Expansion; Toxicity Assay
Quantity 1 Kit