missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies StemRegenin 1 (Hydrochloride)
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- StemRegenin1 (SR1) is an antagonist of the aryl hydrocarbon receptor (AHR)
- It promotes ex vivo expansion of CD34+ human hematopoietic stem cells (Boitano et al.), 2010, Csazar et al.), 2012)
- SR1 has been shown to collaborate with UM729 in preventing differentiation of acute myeloid leukemia (AML) cells in culture (Pabst et al.), 2014)
- SR1 also stimulates the proliferation and differentiation of CD34+ hematopoietic progenitor cells into dendritic cells (Thordardottir et al.), 2014)
- This product is supplied as the hydrochloride salt of the molecule
Specifications
Specifications
| Synonym | SR1 (Hydrochloride) |
| Quantity | 5 mg |
| Molecular Formula | C24H23N5OS · HCl |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction