Learn More
Zeiss™ Stemi 305 Trinocular Stereo Microscope
Product Code. 17322830
Click to view available options
Includes:
Double Spot Illuminator K LED
Ring Illuminator K LED
Ring Illuminator VisiLED
Unit Size:
1 set
Each
Product Code. 17322830
Brand: Zeiss™ 4919030007000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Includes Ring Illuminator K LED
Design Type Trinocular