Learn More
Zeiss™ Stemi 305 Stereo Microscope
Product Code. 15986182
Click to view available options
Includes:
Double Spot Illuminator K LED
Ring Illuminator K LED
Unit Size:
1 set
Each
Product Code. 15986182
Brand: Zeiss™ 4350639020100
This item requires a price update prior to being ordered.
Request a Price for this item here.
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Includes Double Spot Illuminator K LED
Product Type Stereo Microscope
Product Content Correction
Product Title
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado