missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Trilineage Differentiation Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- The STEMdiff™ Trilineage Differentiation Kit provides a simple culture assay to functionally validate the ability of new or established human embryonic stem (ES) and induced pluripotent stem (iPS) cell lines to differentiate to the three germ layers: ectoderm, mesoderm, and endoderm
- This kit includes specialized, complete media and monolayer-based protocols to perform parallel in vitro directed differentiation experiments for each germ layer, clearly and reproducibly establishing trilineage differentiation potential within one week
- STEMdiff™ Trilineage Differentiation Kit is intended to be an endpoint assay and is not optimized for the generation of cells for downstream differentiation or other applications
- STEMdiff™ Trilineage Differentiation Kit has been optimized to assess cells maintained in mTeSR™1
Specifications
Specifications
| Cell Type | Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Characterization, Differentiation, Functional Assay, Phenotyping |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction