Learn More
STEMCELL Technologies STEMdiff™ Neural Rosette Selection Reagent
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- STEMdiff™ Neural Rosette Selection Reagent is an enzyme-free reagent for the selective detachment of neural rosette clusters from adherent neural aggregates previously generated from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells using STEMdiff™ Neural Induction Medium, without manual scraping
- Collecting and re-plating rosette clusters after incubation with STEMdiff™ Neural Rosette Selection Reagent will yield highly pure populations of neural progenitor cells, which can be further sub-cultured as single cells
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Pluripotent Stem Cells |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.