missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Neural Induction Medium

Product Code. 17158301
Encompass
Change view
Click to view available options
Quantity:
250 mL
2 x 250 mL
Unit Size:
250mL
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17158301 2 x 250 mL Pack of 2
17148301 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17158301 Supplier STEMCELL Technologies Supplier No. 05839
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Defined, serum-free medium for neural induction of human ES and iPS cells

  • STEMdiff™ Neural Induction Medium is a defined, serum-free medium for the neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • This medium enables highly efficient generation of neural progenitor cell using either embryoid body- or monolayer culture-based protocols

Specifications

Cell Type Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 2 x 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.