missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Myogenic Progenitor Supplement Kit
Description
- STEMdiff™ Myogenic Progenitor Supplement Kit comprises serum-free supplements for the differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to myogenic progenitor cells
- These cells, which are characterized by myogenic cell markers such as CD56 and CD82, can be culture-expanded for 5+ passages using MyoCult™-SF Expansion Supplement Kit (Human; Catalog #05980) and further differentiated into functional multinucleated MyHC+ myotubes at high efficiency using MyoCult™ Differentiation Kit (Human; Catalog #05965)
- These myotubes can be used in various downstream applications and analyses
- This kit is compatible with human ES and iPS cells maintained in either mTeSR™ 1 (Catalog #85850), mTeSR™ Plus (Catalog #05825), or TeSR™-E8™ (Catalog #05990)
Specifications
Specifications
| Cell Type | Myogenic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?