missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Endothelial Expansion Medium Kit
Description
- STEMdiff™ Endothelial Expansion Medium Kit (Catalog #08007) includes the xeno-free endothelial expansion basal medium and supplement, and must be used in conjunction with Animal Component-Free Cell Attachment Substrate
- STEMdiff™ Endothelial Expansion Medium is optimized for the expansion of hPSC-derived endothelial cells and for the maintenance of endothelial marker expression
- For passaging, Animal Component-Free Cell Dissociation Kit (Catalog #05426) is required
- NOTE: Heparin Solution (Catalog #07980) is required for preparation of complete STEMdiff™ Endothelial Expansion Medium
Specifications
Specifications
| Cell Type | Endothelial Cells |
| For Use With (Application) | Cell Culture; Expansion |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?