Learn More
STEMCELL Technologies STEMdiff™ Cerebral Organoid Kit
Product Code. 16638927
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16638927 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16638927 Supplier STEMCELL Technologies Supplier No. 08570
Culture medium kit for establishment and maturation of human cerebral organoids
  • Generate self-organized, pluripotent stem cell (PSC)-derived neural organoids with a cellular composition and structural organization representative of the developing human brain
  • This simple, four-stage protocol is based on the formulation published by Lancaster et al. (Lancaster MA et al. Nature, 2013 and Lancaster MA et al. Science, 2014) to more reliably generate cerebral organoids
  • Beginning with an embryoid body (EB) formation step followed by expansion of neuroepithelia, organoids generated using STEMdiff Cerebral Organoid Kit feature cortical-like regions including the ventricular zone, outer subventricular zone, intermediate zone, and cortical plate
  • For extended culture periods (> 40 days), the components required for maturation can be purchased as STEMdiff Cerebral Organoid Maturation Kit (Catalog #08571)
Specifications
Cell Type Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation, Organoid Culture
Quantity 1 Kit