missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Cardiomyocyte Support Medium
Description
- STEMdiff™ Cardiomyocyte Support Medium is a versatile medium that limits stress on human pluripotent stem cell (hPSC)-derived cardiomyocytes when transitioning from cryopreservation to thawing and from harvesting to replating of hPSC-derived cardiomyocytes
- After thawing or replating, the functional capacity of hPSC-derived cardiomyocytes is retained and cells can be used in various downstream applications and analyses
- The hPSC-derived cardiomyocytes can be further maintained long-term using STEMdiff™ Cardiomyocyte Maintenance Kit (Catalog #05020)
Specifications
Specifications
| Cell Type | Cardiomyocytes, PSC-Derived |
| For Use With (Application) | Cell Culture |
| Quantity | 250 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?