missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Cardiomyocyte Maintenance Kit

Product Code. 15755778
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15755778 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15755778 Supplier STEMCELL Technologies Supplier No. 05020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium for long-term maintenance of human PSC-derived cardiomyocytes

  • STEMdiff™ Cardiomyocyte Maintenance Kit can be used for long-term maintenance of human pluripotent stem cell (PSC)-derived cardiomyocytes for one month or longer
  • The functional capacity of hPSC-derived cardiomyocytes is retained for downstream applications and analysis
  • This kit is compatible with cardiomyocytes generated using STEMdiff™ Cardiomyocyte Differentiation Kit (Catalog #05010) or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit (Catalog #100-0215)

Specifications

Cell Type Cardiomyocytes, PSC-Derived
For Use With (Application) Cell Culture, Maintenance
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.