missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ APEL™2-LI Medium

Codice prodotto. 17138261
Encompass
Cambia vista
Click to view available options
Quantity:
100 mL
Dimensione della confezione:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
17138261 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17138261 Fornitore STEMCELL Technologies N. del fornitore 05271

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Defined, low-insulin, serum- and animal-component free medium for the differentiation of human ES and iPS cells

  • STEMdiff™ APEL™ 2-LI Medium is a fully defined, low-insulin (LI), serum-free and animal component-free medium for the differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • It is based on the APEL formulation published by Dr
  • Andrew Elefanty and lacks undefined components such as protein-free hybridoma medium
  • STEMdiff™ APEL™ 2-LI can be used in adherent or embryoid body (EB)-based protocols, such as with AggreWell™
  • Appropriate induction factors must be added before use
  • The low insulin content of this medium makes it particularly useful for differentiation to lineages in which insulin is a known inhibitor, such as cardiomyocyte

Specifica

Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 100 mL
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.