Learn More
Cole-Parmer™ Integrity 10, RS-200, and RS-250 Reaction Station Accessories
Product Code. 15447854
Click to view available options
Product Type:
Reducing Sleeve
Reflux Head
Small Volume Test Tube (3 mL)
Small Volume Test Tubes (1 mL)
Unit Size:
Each
Product Code. 15447854
Brand: Cole-Parmer™ 3663087
Keep your reaction station running smoothly with these parts and accessories.
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software
Specifications
Product Type Small Volume Test Tubes (1 mL)
For Use With (Equipment) Integrity 10, RS-200, and RS-250 Reaction Stations
Material Glass