Learn More
Bochem™ Steel Zincked Rod
Product Code. 10222911
Click to view available options
Length (Metric):
500 mm
600 mm
Diameter (Metric) Rod:
12 dia.
12 mm
Unit Size:
Each
Product Code. 10222911
Brand: Bochem™ 5151
Support yourstand bases with the Bochem™ Steel ZIncked Rod, It is sized for M10 stand bases.
  • With thread M10
  • Other sizes available
  • 12 D/O
  • Steel zincked
Specifications
Material Steel Zincked
Length (Metric) 600 mm
Length (Metric) Rod 600 mm
Diameter (Metric) Rod 12 dia.
For Use With (Application) Stand Bases