Learn More
Justrite™ Steel Safety Can for Flammable Liquids
Product Code. 12663135
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12663135 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12663135 Supplier Justrite™ Supplier No. 7110100Z
  • ONLINE EXCLUSIVE PRICE
    84.99€
    63.87€ / Pack of 1
    Save 21.12€
    24% Off
View all promotions
Estimated Shipment: 01-02-2027
Specifications
Certifications/Compliance FM, TUV, UL, ULC, CARB, NFPA, OSHA
Product Type Safety Can
Capacity (Metric) 4 L
Color Red
For Use With (Application) carrying and pouring small quantities of flammable liquids
Material Steel
Finish Powder Coat
Includes sealed lid with automatic positive-pressure relief that vents 0.2 to .35 bar pressure and 40mm fill/pour hole, stainless steel fittings and flame arrester, swinging handle
Diameter (Metric) Outer 241 mm