Learn More
asecos™ Steel Safety Canister with Screw Cap
Product Code. p-200003696 Shop All Asecos Products
Click to view available options
Capacity (Metric):
1 L
2 L
5 L
Unit Size:
Each
Product Code. 17856536
Brand: asecos™ 23129
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
  • With screw cap (secured against loss) overpressure valve, PTFE seal, integrated flame barrier (removable) and plastic protective ring
  • Incl. multilingual safety sticker to designate the contents
Specifications
Product Type Storage Can
Capacity (English) 0.3 gal.
Capacity (Metric) 1 L
Color Yellow
For Use With (Application) Safe storage and transport
Material Steel
Quantity 1
Width (English) 6.50 in.
Width (Metric) 165.00 mm
Height (English) 7.13 in.
Height (Metric) 181.00 mm
Weight (English) 1.59 lb.
Weight (Metric) 0.72 kg