Learn More
Justrite™ Steel Safety Can
Click to view available options
Capacity (Metric):
0.5 L
1 L
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 0.5 L |
| Diameter (Metric) Outer | 122 mm |
| Height (Metric) | 171 mm |