Learn More
Brady™ Maximum Chain Security
Product Code. 15683537
Click to view available options
Diameter (Metric):
8 mm
10 mm
Unit Size:
1 pack
Pack of 2
Product Code. 15683537
Brand: Brady™ 805845
Only null left
Add to basket
Add to basket
More secure than steel cables.
  • Supplied with a fabric sleeve which will protect paintwork and chrome from damage
Specifications
Product Type Chain
Alternate Part Number CHAIN SECURITY 8 MM 2 PK
Diameter (Metric) 8 mm
Quantity 2 Pk.