Learn More
Brady™ Maximum Chain Security
Click to view available options
Diameter (Metric):
8 mm
10 mm
Unit Size:
1 pack
Pack of 2
Description
- Supplied with a fabric sleeve which will protect paintwork and chrome from damage
Specifications
Specifications
| Product Type | Chain |
| Alternate Part Number | CHAIN SECURITY 8 MM 2 PK |
| Diameter (Metric) | 8 mm |
| Quantity | 2 Pk. |